Learn More
Invitrogen™ SNX1 Recombinant Rabbit Monoclonal Antibody (24GB2595)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557733
 View more versions of this product
Product Code. 30275859
  • 198.00 € / 20µL
Description
SNX1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This endosomal protein regulates the cell-surface expression of epidermal growth factor receptor. This protein also has a role in sorting protease-activated receptor-1 from early endosomes to lysosomes. This protein may form oligomeric complexes with family members. This gene results in three transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SNX1
Recombinant Monoclonal
Unconjugated
SNX1
HsT17379; MGC8664; Snx1; SNX1A; sorting nexin 1; sorting nexin 1A; sorting nexin-1; VPS5
Rabbit
Affinity Chromatography
RUO
6642, 84471
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2595
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q13596, Q99N27
SNX1
A synthesized peptide derived from human SNX1 (aa 50-100).
20 μL
Primary
Human, Rat