missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX13 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93194-0.02ml
This item is not returnable.
View return policy
Description
SNX13 Polyclonal antibody specifically detects SNX13 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| SNX13 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| KIAA0713RGS-PX1RGS domain- and PHOX domain-containing protein, sorting nexin 13, sorting nexin-13 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 50-230 of human SNX13 (NP_055947.1). FGKTNSEKYLEQCEHSFLPPTSPGVPKCLEEMKREARTIKIDRRLTGANIIDEPLQQVIQFSLRDYVQYWYYTLSDDESFLLEIRQTLQNALIQFATRSKEIDWQPYFTTRIVDDFGTHLRVFRKAQQKITEKDDQVKGTAEDLVDTFFEVEVEMEKEVCRDLVCTSPKDEEGFLRDLCEV | |
| 0.02 mL | |
| GPCR, Signal Transduction | |
| 23161 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction