missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SNX5 Recombinant Rabbit Monoclonal Antibody (K01_1K91)

Product Code. p-200115186
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30825434 100 μL
30830456 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30825434 Supplier Invitrogen™ Supplier No. MA563180

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: DPSLQIDIPD ALSERDKVKF TVHTKTTLPT FQSPEFSVTR QHEDFVWLHD TLIETTDYAG LIIPPAPTKP DFDGPREKMQ KLGEGEGSMT KEEFAKMKQE Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%, Pig - 100%, Cynomolgus monkey - 100%.

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein binds to fanconi anemia complementation group A protein, but its function is unknown. This gene results in two transcript variants encoding the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX5
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1K91
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene SNX5
Gene Accession No. B1H267, Q9D8U8, Q9Y5X3
Gene Alias 0910001N05Rik; 1810032P22Rik; AU019504; D2Ertd52e; FLJ10931; id:ibd5091; RP11-504H3.2; Snx5; snx5 protein; sorting nexin 5; sorting nexin-5; wu:fa66c10; wu:fi28h04; zgc:55769
Gene Symbols SNX5
Host Species Rabbit
Immunogen Recombinant protein of human SNX5 (aa 28-127).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27131, 296199, 69178
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.