missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOCS-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00 € - 326.00 €
Specifications
| Antigen | SOCS-1 |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 1:300-1:10000 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18681466
|
Novus Biologicals
NBP2-98891-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18613866
|
Novus Biologicals
NBP2-98891-50ul |
50 μg |
216.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SOCS-1 Polyclonal antibody specifically detects SOCS-1 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| SOCS-1 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 8651 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunocytochemistry/ Immunofluorescence 1:300-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| JABTec-interacting protein 3, JAK binding protein, JAK-binding protein, SOCS-1CIS1, SSI-1CISH1, SSI1cytokine-inducible SH2 protein 1, STAT induced SH3 protein 1, STAT-induced STAT inhibitor 1, suppressor of cytokine signaling 1, TIP-3, TIP3Cish1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human SOCS-1. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title