missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PMF1 Antibody (SR2536), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27796
This item is not returnable.
View return policy
Description
PMF1 Monoclonal antibody specifically detects PMF1 in Human,Mouse,Rat samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| PMF1 | |
| Monoclonal | |
| Unconjugated | |
| 50 mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunoprecipitation, Western Blot | |
| SR2536 | |
| Western Blot 1:500-1:1000, Immunoprecipitation 1:20 | |
| PMF-1, Polyamine Modulated Factor 1 | |
| Recombinant protein of human PMF1 (Uniprot #: Q6P1K2) | |
| 100 μL | |
| Cell Biology, Cell Cycle and Replication | |
| 11243 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto