missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOD3/EC-SOD Antibody (SR1423), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00 € - 449.00 €
Specifications
| Antigen | SOD3/EC-SOD |
|---|---|
| Clone | SR1423 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18677514
|
Novus Biologicals
NBP3-21798-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18600825
|
Novus Biologicals
NBP3-21798-50ul |
50 μg |
338.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SOD3/EC-SOD Monoclonal antibody specifically detects SOD3/EC-SOD in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| SOD3/EC-SOD | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC-SODEC 1.15.1.1, extracellular superoxide dismutase [Cu-Zn], MGC20077, superoxide dismutase 3, extracellular | |
| A synthesized peptide derived from human SOD3/EC-SOD (Uniprot #: P08294) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1423 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cancer, Neurodegeneration, Neuroscience | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 6649 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title