missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-pentanesulfonate monohydrate, HPLC grade

Product Code. 11397128
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397128 100 g 100g
11317138 5 g 5g
11307138 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11397128 Supplier Thermo Scientific Alfa Aesar Supplier No. 015714.22
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ion pair reagent | CAS: 207605-40-1 | C5H13NaO4S | 192.205 g/mol

Ion pair reagent.Sodium 1-pentanesulfonate monohydrate is used as a surfactant and a reversed-phase ion-pairing agent. It is utilized to study its effects on the properties of polyaniline, using Fourier transform infrared and UV-visible spectroscopy. Further, it is used for HPLC analysis of proteins and peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207605-40-1
Molecular Formula C5H13NaO4S
Molecular Weight (g/mol) 192.205
MDL Number MFCD00007541
InChI Key FPQYXAFKHLSWTI-UHFFFAOYSA-M
Synonym sodium pentane-1-sulfonate hydrate, 1-pentanesulfonic acid sodium salt monohydrate, sodium 1-pentanesulfonate monohydrate, potassium hydrate pentane-1-sulfonate, sodium 1-pentanesulfonate hydrate, sodium hydrate pentane-1-sulfonate, sodium pentane-1-sulfonate-water 1/1/1, 1-pentane sulphonic acid sodium salt monohydrate, 1-pentanesulfonic acid sodium salt monohydydrate, sodium 1-pentanesulfonate monohydrate, hplc grade
PubChem CID 23693099
IUPAC Name sodium;pentane-1-sulfonate;hydrate
SMILES CCCCCS(=O)(=O)[O-].O.[Na+]

Specifications

Color White
Linear Formula CH3(CH2)4SO3Na·H2O
Quantity 100 g
Beilstein 3725147
Solubility Information Soluble in water.
Formula Weight 192.21 (174.19 Anhydrous)
Grade HPLC
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-pentanesulfonate monohydrate
compliance-icons
Product Identifier
  • Sodium 1-pentanesulfonate monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Sodium 1-pentanesulfonate monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.