missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-bromoethanesulfonate, 98%

Product Code. 11427377
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11427377 25 g 25g
11437377 100 g 100g
11447377 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11427377 Supplier Thermo Scientific Alfa Aesar Supplier No. A15076.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4263-52-9 | C2H4BrNaO3S | 211.005 g/mol

Sodium 2-bromoethanesulfonate(BES) was used to investigate the effect of BES on inhibition of bacterial growth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 2-bromoethanesulfonate(BES) was used to investigate the effect of BES on inhibition of bacterial growth.

Solubility
Soluble in water and methanol.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4263-52-9
Molecular Formula C2H4BrNaO3S
Molecular Weight (g/mol) 211.005
MDL Number MFCD00007530
InChI Key HNFOAHXBHLWKNF-UHFFFAOYSA-M
Synonym sodium 2-bromoethanesulfonate, 2-bromoethanesulfonic acid sodium salt, sodium 2-bromoethanesulphonate, 2-bromoethanesulfonic acid, sodium salt, sodium bromoethylsulfonate, ethanesulfonic acid, 2-bromo-, sodium salt, ethanesulfonic acid, 2-bromo-, sodium salt 1:1, sodium 2-bromoethane sulfonate, 2-bromoethanesulfonic acid sodium, natrium-2-bromethansulfonat
PubChem CID 23666797
IUPAC Name sodium;2-bromoethanesulfonate
SMILES C(CBr)S(=O)(=O)[O-].[Na+]

Specifications

Melting Point ∼284°C (decomposition)
Quantity 25 g
Beilstein 4590828
Solubility Information Soluble in water and methanol.
Formula Weight 211.02
Percent Purity 98%
Chemical Name or Material Sodium 2-bromoethanesulfonate
compliance-icons
Product Identifier
  • 2-Bromoethanesulfonic acid, sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Bromoethanesulfonic acid, sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.