missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-Ethylhexanoate, TRC

Product Code. 30414223
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30414223 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30414223 Supplier Toronto Research Chemicals Supplier No. TRCS6240805G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19766-89-3
Molecular Formula C8H15NaO2
Molecular Weight (g/mol) 166.19
Synonym 2-Ethylhexanoic Acid Sodium Salt, 2-Ethylcaproic Acid Sodium Salt, Sodium 2-Ethylcaproate
IUPAC Name sodium;2-ethylhexanoate
SMILES [Na+].CCCCC(CC)C(=O)[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H16O2.Na/c1-3-5-6-7(4-2)8(9)10;/h7H,3-6H2,1-2H3,(H,9,10);/q;+1/p-1
Formula Weight 166.097
Chemical Name or Material Sodium 2-Ethylhexanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.