missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3-nitrobenzoate, 94%, may contain up to ca 10% water

Product Code. 11312407
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11312407 500 g 500g
11302407 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312407 Supplier Thermo Scientific Alfa Aesar Supplier No. L10028.36
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 827-95-2 | C7H4NNaO4 | 189.10 g/mol

Sodium 3-nitrobenzoate is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 3-nitrobenzoate is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 827-95-2
Molecular Formula C7H4NNaO4
Molecular Weight (g/mol) 189.10
MDL Number MFCD00150535,MFCD00051097
InChI Key MUADFEZFSKAZLT-UHFFFAOYSA-M
Synonym sodium 3-nitrobenzoate, 3-nitrobenzoic acid sodium salt, benzoic acid, 3-nitro-, sodium salt, unii-i1eq837b3j, m-nitrobenzoic acid sodium salt, sodium m-nitrobenzoate, benzoic acid, 3-nitro-, sodium salt 1:1, 3-nitrobenzoic acid, sodium salt, acmc-209pqe, dsstox_cid_24733
PubChem CID 70014
IUPAC Name sodium;3-nitrobenzoate
SMILES [Na+].[O-]C(=O)C1=CC=CC(=C1)[N+]([O-])=O

Specifications

Melting Point >300°C
Quantity 500 g
Beilstein 4166479
Solubility Information Soluble in water.
Formula Weight 189.1
Percent Purity 94%
Assay may contain up to ≈10% water
Chemical Name or Material Sodium 3-nitrobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.