missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-aminosalicylate dihydrate, MedChemExpress

Product Code. 16491238
Encompass_Preferred
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
missing translation for 'unitSize':
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
16491238 5 g 5g
16472459 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 16491238 missing translation for 'mfr' Medchem Express missing translation for 'supplierNo' HYI0447A
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sodium 4-aminosalicylate dihydrate (4-Aminosalicylic acid sodium salt dihydrate) is one of the antimycobacterial drugs currently used for multidrug-resistant tuberculosis.

Sodium 4-aminosalicylate dihydrate (4-Aminosalicylic acid sodium salt dihydrate) is one of the antimycobacterial drugs currently used for multidrug-resistant tuberculosis.

Specifications

Chemical Name or Material Sodium 4-aminosalicylate dihydrate
CAS 6018-19-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C7H10NNaO5
Quantity 5 g
Synonym 4-Aminosalicylic acid sodium salt dihydrate
Solubility Information H2O : 100 mg/mL (473.60 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (473.60 mM; Need ultrasonic)
SMILES O=C(O[Na])C1=C(O)C=C(N)C=C1.O.O
Molecular Weight (g/mol) 211.15
Formula Weight 211.15
Percent Purity 99.78%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.