missing translation for 'onlineSavingsMsg'
Learn More

Sodium for AAS 1g/L H2O matrix, Fisher chemical™
BIOPHARMA

Product Code. 12953305 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12953305 500 mL Bottle 500mL
12943305 100 mL Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12953305 Supplier Fisher Chemical Supplier No. B 235 500
  • ONLINE EXCLUSIVE PRICE
    60.00 €
    46.17 € / 500mL
    Save 13.83 €
    23% Off
 Restricted Item
Estimated Shipment: 28-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Chemical Identifiers

CAS 7440-23-5
Molecular Formula Na
Molecular Weight (g/mol) 22.99
MDL Number MFCD00085307
InChI Key KEAYESYHFKHZAL-UHFFFAOYSA-N
PubChem CID 5360545
ChEBI CHEBI:26708
IUPAC Name sodium
SMILES [Na]

Specifications

Packaging Bottle
Physical Form Liquid
Concentration 1 g/L ±0.002 g/L
Type Flame Single-element Standard Solution
Solvent or Matrix Water
Quantity 500 mL
Chemical Name or Material Sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.