missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate, 1.00 M (1.00 N), Ricca Chemical

Product Code. 31750352
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31750352 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31750352 Supplier Ricca Chemical Company Supplier No. R71295001A
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 6131-90-4, 7732-18-5
CAS Min % 86.0, 11.75
CAS Max % 90.0, 12.22
Molecular Formula C2H9NaO5
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
SMILES CC(=O)[O-].O.O.O.[Na+]
IUPAC Name sodium;acetate;trihydrate
Molecular Weight (g/mol) 136.079
PubChem CID 23665404
ChEBI CHEBI:32138
Grade Laboratory
Packaging Natural Poly Bottle
pH 7.5 to 9.2
Quantity 1 L
Concentration or Composition Notes 1.00Molar,1.00Normal
Chemical Name or Material Sodium Acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.