missing translation for 'onlineSavingsMsg'
Learn More

Sodium bicarbonate, 1M buffer soln., pH 8.5

Product Code. 15483179
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15483179 500 mL 500mL
15473179 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15483179 Supplier Thermo Scientific Alfa Aesar Supplier No. J60408.AP
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 144-55-8 | CHNaO3 | 84.01 g/mol

Sodium bicarbonate acts a weak buffer with that makes maintaining a pH range of 7.2 - 7.6. It is also used as a nutrient source. This solution prepared in water is used as injection in Pharmaceutical industry.is used to treat cement contamination ins hore pac drilling slurry and bentonite water based drilling fluids.It is also used as a buffer. Sodium bicarbonate will remove calcium and manganese making hard water soft.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium bicarbonate acts a weak buffer with that makes maintaining a pH range of 7.2 - 7.6. It is also used as a nutrient source. This solution prepared in water is used as injection in Pharmaceutical industry.is used to treat cement contamination ins hore pac drilling slurry and bentonite water based drilling fluids.It is also used as a buffer. Sodium bicarbonate will remove calcium and manganese making hard water soft.

Solubility
Soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away fom strong acids, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium;hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Concentration 1 M
pH 8.5
Physical Form Liquid
Quantity 500 mL
Solubility Information Soluble in water
Chemical Name or Material Sodium bicarbonate buffer soln.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.