missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium bicarbonate, 1M buffer soln., pH 9.4

Product Code. 15455109
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15455109 250 mL 250mL
15465109 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455109 Supplier Thermo Scientific Chemicals Supplier No. J62808.AK
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 144-55-8 | CHNaO3 | 84.01 g/mol

The bicarbonate buffering system is important in many different cellular processes. It is also used in the formation of acid in the lumen on the stomach. Used to neutralize the pH of the chyme leaving the stomach and entering the small intestine. It is also used to maintain pH in serum samples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The bicarbonate buffering system is important in many different cellular processes. It is also used in the formation of acid in the lumen on the stomach. Used to neutralize the pH of the chyme leaving the stomach and entering the small intestine. It is also used to maintain pH in serum samples.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium;hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Concentration 1 M
pH 9.4
Physical Form Liquid
Quantity 250 mL
Solubility Information It is soluble in water.
Chemical Name or Material Sodium bicarbonate buffer soln.
compliance-icons
Product Identifier
  • Sodium bicarbonate, 1M buffer solution, pH 9.4
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Disodium carbonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.