missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Calcium Exchanger 1/NCX1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94316-0.1ml
This item is not returnable.
View return policy
Description
Sodium Calcium Exchanger 1/NCX1 Polyclonal antibody specifically detects Sodium Calcium Exchanger 1/NCX1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Sodium Calcium Exchanger 1/NCX1 | |
| Polyclonal | |
| Western Blot 1:2000 - 1:6000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| FLJ37694, FLJ43417, Na(+)/Ca(2+)-exchange protein 1, sodium/calcium exchanger 1, solute carrier family 8 (sodium/calcium exchanger), member 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 600-700 of human Sodium Calcium Exchanger 1/NCX1 (NP_066920.1). DEIVKTISVKVIDDEEYEKNKTFFLEIGEPRLVEMSEKKALLLNELGGFTITGKYLFGQPVFRKVHAREHPILSTVITIADEYDDKQPLTSKEEEERRIAE | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Endocrinology, Neuroscience, Signal Transduction | |
| 6546 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction