missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Analyte or Component Names | Sodium Carbonate R1 |
| Purity Grade Notes | EU Ph Ref: 1079302 |
| Element | Sodium Carbonate R1 |
| Grade | Eu. Ph. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction
