missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Sodium Chloride (NaCl) Solutions, 0.1–2.0 M

Product Code. 18014100 Shop All Gibco Products
Change view
Click to view available options
Quantity:
5 L
20 L
200 L
Packaging:
Drum
Sample
Tote
Concentration:
0.1 M
0.2 M
0.5 M
1.0 M
2.0 M
Unit Size:
20 L
200 L
5 L
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Concentration Unit Size
18014100 20 L Tote 0.1 M 20 L
18014060 5 L Sample 0.1 M 5 L
18024070 200 L Drum 0.1 M 200 L
18024100 5 L Sample 0.2 M 5 L
18024080 20 L Tote 0.2 M 20 L
18054080 200 L Drum 0.2 M 200 L
18064100 5 L Sample 0.5 M 5 L
18074070 20 L Tote 0.5 M 20 L
18064080 200 L Drum 0.5 M 200 L
18094080 5 L Sample 1.0 M 5 L
18074080 20 L Tote 1.0 M 20 L
18024060 200 L Drum 1.0 M 200 L
18004070 5 L Sample 2.0 M 5 L
18014090 20 L Tote 2.0 M 20 L
18084070 200 L Drum 2.0 M 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Product Code. 18014100 Supplier Gibco™ Supplier No. A5792202
  • ONLINE EXCLUSIVE PRICE
    899.00 €
    689.98 € / 20 L
    Save 209.02 €
    23% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gibco Sodium Chloride (NaCl) solutions are sterile-filtered, packaged in bioprocess containers, and ready-to-use. Sourcing NaCl solutions can help streamline your buffer preparation allowing you to focus on additional critical steps in your biopharmaceutical processes.

We mix compendial-grade sodium chloride (NaCl) with the same bulk water used in our Gibco Water for Injection (WFI) for Cell Culture to produce solutions that meet the most stringent expectations of the pharmaceutical, biopharma, and diagnostic industries. NaCl solutions are manufactured to cGMP (21CFR820) and ISO 13485 standards, promoting consistency and convenience for every stage of the bioprocessing pipeline.

Product features:
• Multiple concentrations available (0.1 M, 0.2 M, 0.5 M, 1.0 M, 2.0 M)
• Available in a range of pack sizes (5 L, 20 L, 200 L)
• Established products with set specifications are ready to order
• Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
• Readily accessible documentation for accelerated qualification

User benefits:
• Eases the time requirements dedicated for in-house buffer preparation
• Mitigates space required for equipment storage
• Streamlines in-house chromatography processes related to formulation, hydration, testing, and validation
• Meets workflow needs for all stages of biologic development and manufacturing

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Chloride
CAS 7647-14-5
Color Clear
Concentration 0.1 M
For Use With (Application) Bioproduction
pH 4.5–8.0
Physical Form Liquid
Quantity 20 L
Solution Type Buffer
Formulation 0.1 M NaCl
Packaging Tote
Sterility Sterile-filtered
Certifications/Compliance GMP - 21 CFR 820, ISO 13485
Sterilization Method 0.2 μm Sterile-filtered
Endotoxin Level ≤ 1.0 EU/mL
Osmolality 167–207 mOsm/kg
Molecular Weight (g/mol) 58.44
Grade Compendial
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.