missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride Solution, 30g/L, Chem-Lab

Product Code. 11900725
Click to view available options
Quantity:
250 mL
missing translation for 'unitSize'
Each
This item is not returnable. View return policy

Product Code. 11900725

missing translation for 'mfr': Chem-Lab CL03.1414.0250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for salinity measurement. Sodium Chloride Solution, 30g/L, Chem-Lab is an electrolyte solution, also known as saline. This standard reference solution is used in salinity determination, in equipment calibration and as a chemical reagent in many laboratory applications.

This sodium chloride standard solution contains a precise concentration of 30g/L of NaCl per liter of water. It is used to verify the accuracy of salinity determinations and to calibrate equipment used in those determinations.

Specifications

Packaging Polyethylene
CAS 7647-14-5, 7732-18-5
Odor Odorless
Density 1.02g/mL
Physical Form Liquid
Molecular Formula ClNa
Concentration 30 g/L
MDL Number MFCD00003477
Color Colorless
pH 6 to 8
Synonym Saline
Solubility Soluble
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium;chloride
Molecular Weight (g/mol) 58.44
PubChem CID 5234
Quantity 250 mL
ChEBI CHEBI:26710
Formula Weight 58.44g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.