missing translation for 'onlineSavingsMsg'
Learn More

Sodium Deoxycholate, TRC

Product Code. 30420644
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30420644 5 g
30416577 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30420644 Supplier Toronto Research Chemicals Supplier No. TRCS6240905G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 302-95-4
Molecular Formula C24 H39 O4 . Na
Molecular Weight (g/mol) 414.5538
Synonym Cholan-24-oic acid, 3,12-dihydroxy-, sodium salt (1:1), (3α,5β,12α)-, 5β-Cholan-24-oic acid, 3α,12α-dihydroxy-, monosodium salt (8CI), Cholan-24-oic acid, 3,12-dihydroxy-, monosodium salt, (3α,5β,12α)- (9CI), Deoxycholic acid monosodium salt, Deoxycholic acid sodium salt, Desoxycholate sodium, Desoxycholic acid sodium salt, NSC 681065, Sodium 3α,12α-dihydroxy-5β-cholanate, Sodium 7-deoxycholate, Sodium deoxycholate, Sodium desoxycholate
IUPAC Name sodium;(4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
SMILES [Na+].C[C@H](CCC(=O)[O-])[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C24H40O4.Na/c1-14(4-9-22(27)28)18-7-8-19-17-6-5-15-12-16(25)10-11-23(15,2)20(17)13-21(26)24(18,19)3;/h14-21,25-26H,4-13H2,1-3H3,(H,27,28);/q;+1/p-1/t14-,15-,16-,17+,18-,19+,20+,21+,23+,24-;/m1./s1
Formula Weight 414.2746
Chemical Name or Material Sodium Deoxycholate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.