accessibility menu, dialog, popup

UserName

CAS RN 302-95-4

IUPAC Name: sodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate
Synonyms: sodium deoxycholate deoxycholic acid sodium salt sodium 7-deoxycholate sodium desoxycholate desoxycholate sodium deoxycholate sodium
Molecular Weight (g/mol): 414.56
Molecular Formula: C24H39NaO4
InChi Key: FHHPUSMSKHSNKW-SMOYURAASA-M
SMILES: [Na+].C[C@H](CCC([O-])=O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)