missing translation for 'onlineSavingsMsg'
Learn More

Sodium ethoxide, pure, 21% in ethanol

Product Code. p-192462 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
250g
This item is not returnable. View return policy

Product Code. 10570322

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 141-52-6 | C2H5NaO | 68.051 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-52-6, 64-17-5
Molecular Formula C2H5NaO
Molecular Weight (g/mol) 68.051
MDL Number MFCD00012417
InChI Key QDRKDTQENPPHOJ-UHFFFAOYSA-N
Synonym sodium ethoxide, sodium ethylate, sodium ethanolate, sodiumethoxide, ethoxysodium, ethanol, sodium salt, caustic alcohol, naoet, etona, ethanol, sodium salt 1:1
PubChem CID 2723922
ChEBI CHEBI:52096
IUPAC Name sodium;ethanolate
SMILES CC[O-].[Na+]

Specifications

Color Yellow to Brown
Boiling Point 91.0°C
Linear Formula C2H5ONa
Beilstein 01, 311
Fieser 01,1065; 03,265; 04,451; 06,540
Density 0.8680g/mL
Merck Index 15, 8749
Solubility Information Solubility in water: miscible (hydrolyse).
Formula Weight 68.04
Percent Purity 20.5 to 21.5% (NaOC2H5)
Grade Pure
Packaging Glass bottle
Refractive Index 1.385
Specific Gravity 0.868
Viscosity 23 mPa.s (20°C)
Quantity 100 g
Flash Point 25°C
Physical Form Liquid
Chemical Name or Material Sodium ethoxide, 21% in Ethanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium ethoxide, 21% in ethanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

For Manufacturing and Laboratory Use. Not for beverage use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.