missing translation for 'onlineSavingsMsg'
Learn More

Sodium hippurate, 96%

Product Code. p-7081659
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11429243

Brand: Thermo Scientific Alfa Aesar A11723.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 532-94-5 | C9H8NNaO3 | 201.157 g/mol

It may be a useful urinary biomarker for certain cancers such as gastric cancer and oxidative stress.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It may be a useful urinary biomarker for certain cancers such as gastric cancer and oxidative stress.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532-94-5
Molecular Formula C9H8NNaO3
Molecular Weight (g/mol) 201.157
MDL Number MFCD00002693
InChI Key ZBCAZEFVTIBZJS-UHFFFAOYSA-M
Synonym sodium hippurate, hippuric acid sodium salt, sodium 2-benzamidoacetate, n-benzoylglycine sodium salt, glycine, n-benzoyl-, monosodium salt, ccris 5812, hippuric acid, monosodium salt, glycine, n-benzoyl-, sodium salt 1:1, hippurate sodium, benzoylaminoacetic acid sodium salt
PubChem CID 516953
IUPAC Name sodium;2-benzamidoacetate
SMILES C1=CC=C(C=C1)C(=O)NCC(=O)[O-].[Na+]

Specifications

Assay Percent Range 96%
Quantity 50 g
Solubility Information Soluble in water.
Formula Weight 201.16
Percent Purity 96%
Physical Form Powder or Lumps
Chemical Name or Material Sodium Hippurate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.