missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Rhesus Macaque TNF-alpha Protein

Product Code. 18606535 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18606535 500 μg 500µg
18673956 5 μg 5µg
18688215 1 mg 1mg
18693246 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18606535 Supplier R&D Systems™ Supplier No. NBP235321500UG
  • ONLINE EXCLUSIVE PRICE
    3140.95 €
    2789.85 € / 500µg
    Save 351.10 €
    11% Off
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Recombinant Protein

TNF alpha Protein is fully biologically active when compared to standard. The ED50 as determined by a cytotoxicity assay using murine L929 cells is less than 0.05 ng/mL, corresponding to a specific activity of > 2.0 x 10^7 IU/mg in the presence of actinomycin D.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P48094, P48094, P48094
For Use With (Application) SDS-PAGE, Bioactivity
Formulation Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4.
Gene ID (Entrez) 7124
Molecular Weight (g/mol) MolecularWeight-theroretical: 17.3 kDa
Quantity 500 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin Concentration Less than 1 EU/μg of TNF-alpha as determined by LAL method.
Gene Alias APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alpha, TNF-alpha/TNFSF2, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Gene Symbol TNF
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Research Category Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
Product Line Novus
Purity or Quality Grade >95%, by SDS-PAGE and HPLC
Protein TNF-alpha
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.