missing translation for 'onlineSavingsMsg'
Learn More

Sodium methoxide, pure, titrant, (0.5 M in methanol)

Product Code. 11965871 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11965871 2.5 L Glass bottle 2.5 L
10458643 100 mL Glass bottle 100mL
10637391 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11965871 Supplier Thermo Scientific Acros Supplier No. 419600025
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 124-41-4 | CH3NaO | 54.024 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-41-4 , 67-56-1
Molecular Formula CH3NaO
Molecular Weight (g/mol) 54.024
MDL Number MFCD00012179
InChI Key WQDUMFSSJAZKTM-UHFFFAOYSA-N
Synonym sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
PubChem CID 10942334
IUPAC Name sodium;methanolate
SMILES C[O-].[Na+]

Specifications

Color Colorless
Boiling Point 65.0°C
Melting Point -98.0°C
Linear Formula CH3ONa
Concentration 0.45 to 0.60M
Fieser 01,1091; 02,385; 03,259; 04,457; 05,617; 06,545; 08,463; 11,489; 13,352
Density 0.8000g/mL
Solubility Information Solubility in water: reacts.
Formula Weight 54.02
Grade Pure
Packaging Glass bottle
Specific Gravity 0.8
Quantity 2.5 L
Flash Point 11°C
Physical Form Liquid
Chemical Name or Material Sodium methoxide, 0.5 M in methanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.