missing translation for 'onlineSavingsMsg'
Learn More

Sodium octanoate, 96%

Product Code. 11485513
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485513 500 g 500g
11475513 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11485513 Supplier Thermo Scientific Alfa Aesar Supplier No. A13451.36
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1984-06-1 | C8H15NaO2 | 166.20 g/mol

Sodium octanoate may be used as a protein stabilizer for molecules such as albumin and fibrinogen under conditions such heat sterilization. Also used for blood plasma fractionation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium octanoate may be used as a protein stabilizer for molecules such as albumin and fibrinogen under conditions such heat sterilization. Also used for blood plasma fractionation.

Solubility
Soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1984-06-1
MDL Number MFCD00058511

Specifications

Melting Point >300°C
Molecular Formula C8H15NaO2
Quantity 500 g
Beilstein 3597723
Synonym sodium octanoate, sodium caprylate, octanoic acid, sodium salt, sodium n-octanoate, sodium octoate, caprylic acid sodium salt, natrium octanoat, octanoic acid sodium salt, unii-9xtm81vk2b, 9xtm81vk2b
Solubility Information Soluble in water.
InChI Key BYKRNSHANADUFY-UHFFFAOYSA-M
SMILES CCCCCCCC(=O)[O-].[Na+]
IUPAC Name sodium;octanoate
Molecular Weight (g/mol) 166.20
PubChem CID 23664772
Formula Weight 166.2
Percent Purity 96%
Chemical Name or Material Sodium octanoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.