missing translation for 'onlineSavingsMsg'
Learn More

Sodium oxalate, 0.1N Standardized Solution

Product Code. 15452857
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15452857 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15452857 Supplier Thermo Scientific Alfa Aesar Supplier No. 047123.K2
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 62-76-0 | C2Na2O4 | 134.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 62-76-0
Molecular Formula C2Na2O4
Linear Formula Na2C2O4
MDL Number MFCD00012465
Quantity 1 L
Physical Form Liquid
Synonym sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O
IUPAC Name disodium oxalate
Molecular Weight (g/mol) 134.00
PubChem CID 6125
Formula Weight 134
Chemical Name or Material Sodium oxalate, 0.1N Standardized Solution
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.