missing translation for 'onlineSavingsMsg'
Learn More

Sodium phenylpyruvate, 98%

Product Code. 15413308
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15413308 5 g 5g
15423308 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15413308 Supplier Thermo Scientific Alfa Aesar Supplier No. H56767.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 114-76-1 | C9H7NaO3 | 186.142 g/mol

Phenylpyruvic acid, sodium salt is a substrate for phenylpyruvate decarboxylase and phenylpyruvate tautomerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylpyruvic acid, sodium salt is a substrate for phenylpyruvate decarboxylase and phenylpyruvate tautomerase.

Solubility
Soluble in water

Notes
Store at 2-8°C. Avoin oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114-76-1
Molecular Formula C9H7NaO3
Molecular Weight (g/mol) 186.142
MDL Number MFCD00002590
InChI Key MQGYVGKMCRDEAF-UHFFFAOYSA-M
Synonym sodium phenylpyruvate, phenylpyruvic acid sodium salt, sodium 2-oxo-3-phenylpropanoate, unii-7528cw33ww, sodium ?-phenylpyruvate, phenylpyruvic acid, sodium salt, l-phenyl pyruvic acid sodium salt, beta-phenylpyruvic acid sodium salt, 2-oxo-3-phenylpropionic acid, 3-phenyl-2-oxopropanoic acid, sodium salt
PubChem CID 23667645
IUPAC Name sodium;2-oxo-3-phenylpropanoate
SMILES C1=CC=C(C=C1)CC(=O)C(=O)[O-].[Na+]

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 4770978
Solubility Information Soluble in water
Formula Weight 186.14
Percent Purity 98%
Chemical Name or Material Sodium phenylpyruvate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.