missing translation for 'onlineSavingsMsg'
Learn More

Sodium phenylpyruvate monohydrate, 98%

Product Code. 11457906
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
50 g
Unit Size:
1g
25g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457906 1 g 1g
11467906 5 g 5g
11477906 25 g 25g
11464030 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11457906 Supplier Thermo Scientific Alfa Aesar Supplier No. A14027.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 122049-54-1 | C9H7O3 | 163.15 g/mol

Sodium phenylpyruvate monohydrate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium phenylpyruvate monohydrate is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122049-54-1
Molecular Formula C9H7O3
Molecular Weight (g/mol) 163.15
MDL Number MFCD00149109
InChI Key BTNMPGBKDVTSJY-UHFFFAOYSA-M
Synonym sodium phenylpyruvate monohydrate, sodium 2-oxo-3-phenylpropanoate hydrate, phenylpyruvic acid sodium salt monohydrate, phenylpyruvic acid, sodium salt, sodium hydrate ?-phenylpyruvate, l-phenyl pyruvic acid sodium salt, beta-phenylpyruvic acid sodium salt, acmc-2099lh, phenylpyruvic acid sodium hydrate, beta-phenylpyruvic acid, sodium salt
PubChem CID 23666336
IUPAC Name sodium;2-oxo-3-phenylpropanoate;hydrate
SMILES [O-]C(=O)C(=O)CC1=CC=CC=C1

Specifications

Melting Point >300°C
Linear Formula C6H5CH2COCO2Na·H2O
Quantity 1 g
Beilstein 4770978
Solubility Information Soluble in water.
Formula Weight 204.16 (186.14 Anhydrous)
Percent Purity 98%
Chemical Name or Material Sodium phenylpyruvate monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Politique de confidentialité.