missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium phosphate, 0.2M buffer soln., pH 7.0

Product Code. 15425709
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15425709 500 mL 500mL
15435709 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15425709 Supplier Thermo Scientific Chemicals Supplier No. J63482.AP
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium phosphate is widely used as a reagent with high buffering capacity for molecular biology, biochemistry, and chromatography

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium phosphate is widely used as a reagent with high buffering capacity for molecular biology, biochemistry, and chromatography

Solubility
Fully miscible.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium phosphate buffer soln.
Concentration 0.2 M
pH 7.0
Physical Form Liquid
Quantity 500 mL
MDL Number MFCD00146206
Solubility Information Fully miscible.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.