Learn More
Thermo Scientific Chemicals Sodium phosphate, 0.5M buffer soln., pH 8.5
Brand: Thermo Scientific Chemicals J61151.AK
Code : NEW
Additional Details : Weight : 0.00025kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium phosphate buffer soln. | |
8.5 | |
250 mL |
0.5 M | |
Liquid | |
MFCD00146206 |
RUO – Research Use Only