missing translation for 'onlineSavingsMsg'
Learn More

Sodium Salicylate, BAKER ANALYZED, J.T. Baker™

Product Code. 10092520 Shop All J.T.Baker Products
Change view
Click to view available options
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10092520 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10092520 Supplier JT Baker Supplier No. 2028.0250
Quantity Price
1 52.49 € / 250g
10 48.20 € / 250g

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Laboratory Reagent

Specifications

CAS 54-21-7
Melting Point 200°C
Assay Percent Range min. 99.5%
Molecular Formula C7H5NaO3
MDL Number MFCD00002440
Synonym Salicylate de sodium
InChI Key ABBQHOQBGMUPJH-UHFFFAOYSA-M
SMILES [Na+].OC1=CC=CC=C1C([O-])=O
IUPAC Name sodium 2-hydroxybenzoate
Molecular Weight (g/mol) 160.10
Density 0.32 g/cm³ (20°C)
Quantity 250 g
Assay After drying
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.