missing translation for 'onlineSavingsMsg'
Learn More

Sodium Salicylate, MP Biomedicals

Product Code. 11438755 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438755 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11438755 Supplier MP Biomedicals Supplier No. 0522476201
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 54-21-7
Molecular Formula C7H5NaO3
Quantity 1 kg
Synonym sodium salicylate, sodium 2-hydroxybenzoate, salsonin, clin, enterosalicyl, enterosalil, entrosalyl, glutosalyl, kerasalicyl, magsalyl
InChI Key ABBQHOQBGMUPJH-UHFFFAOYSA-M
SMILES C1=CC=C(C(=C1)C(=O)[O-])O.[Na+]
IUPAC Name sodium;2-hydroxybenzoate
Molecular Weight (g/mol) 160.104
PubChem CID 16760658
ChEBI CHEBI:9180
Formula Weight 160.11
Chemical Name or Material Sodium Salicylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.