missing translation for 'onlineSavingsMsg'
Learn More

Sodium Salicylate, TRC

Product Code. 30009957
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
100g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009957 500 g 500g
30012635 250 g 250g
30006094 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009957 Supplier Toronto Research Chemicals Supplier No. TRCS634900500G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-21-7
Molecular Formula C7 H5 O3 . Na
Molecular Weight (g/mol) 160.1
Synonym Benzoic acid, 2-hydroxy-, monosodium salt (9CI), Salicylic acid, monosodium salt (8CI), Sodium salicylate (7CI), Alysine, Aroall, Camporisin, Clin, Enterosalicyl, Enterosalil, Entrosalyl, Glutosalyl, Idocyl, Kerasalicyl, Kerosal, Magsalyl, Monosodium salicylate, Nadisal, Neo-Salicyl, Salicylic acid sodium salt, Saliject, Salisod, Salsonin, Sodium 2-hydroxybenzoate, Sodium o-hydroxybenzoate, Sodium salicylate (NaO3C7H5), o-Hydroxybenzoic acid monosodium salt
IUPAC Name sodium;2-hydroxybenzoate
SMILES [Na+].Oc1ccccc1C(=O)[O-]

Specifications

Quantity 500 g
InChI Formula InChI=1S/C7H6O3.Na/c8-6-4-2-1-3-5(6)7(9)10;/h1-4,8H,(H,9,10);/q;+1/p-1
Formula Weight 160.0136
Chemical Name or Material Sodium Salicylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.