accessibility menu, dialog, popup

UserName

NaOHOO

CAS RN 54-21-7

IUPAC Name: sodium 2-hydroxybenzoate
Synonyms: salicylate-sodium
Molecular Weight (g/mol): 160.1
Molecular Formula: C7H5NaO3
InChi Key: ABBQHOQBGMUPJH-UHFFFAOYSA-M
SMILES: [Na+].OC1=CC=CC=C1C([O-])=O
Molecular Weight (g/mol) 
  • (7)
  • (1)
  • (8)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (3)
Percent Purity 
  • (4)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (4)
  • (3)
Grade 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (2)
  • (7)
  • (1)
  • (8)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (2)
  • (4)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)