missing translation for 'onlineSavingsMsg'
Learn More

Sodium Standard: Na+ @ 1000 μg/mL in H2O, VHG™

Product Code. 17957844
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17957844 500 mL 500mL
17968234 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17957844 Supplier VHG Labs Supplier No. VHGINAW1K500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Sodium Standard: Na+ @ 1000 μg/mL in H2O
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium
CAS 7440-23-5
Methods Ion Chromatography
Analyte or Component Names Sodium
Shipping Condition Room Temperature
Molecular Formula Na
Certifications/Compliance ISO 17034
Synonym Natrium, Sodio, Sodium metal
InChI Formula InChI=1S/Na
SMILES [Na]
IUPAC Name sodium
Solvent or Matrix Water
Molecular Weight (g/mol) 22.99
Quantity 500 mL
Formula Weight 22.9898 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.