missing translation for 'onlineSavingsMsg'
Learn More

Sodium Standard For ICP/MS, Claritas PPT Grade, SPEX CertiPrep™

Product Code. 11445998 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445998 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11445998 Supplier SPEX Certiprep Supplier No. CLNA22Y
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Na1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging Triple-leached LDPE bottle
CAS 7440-23-5, 7732-18-5, 7732-18-5
Certificate of Analysis Claritas PPT™ Certificate of Analysis reports up to 68 elements at ppt levels.
CAS Min % 0.1, 2.0, 97.9
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 0.1, 2.0, 97.9
Molecular Formula Na
MDL Number MFCD00085307
Technique ICP-MS
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solution Type Standard Solution
InChI Key KEAYESYHFKHZAL-UHFFFAOYSA-N
Type ICP/ICP-MS Standard
SMILES [Na]
IUPAC Name sodium
Solvent or Matrix 2% HNO3
Molecular Weight (g/mol) 22.99
PubChem CID 5360545
Quantity 125 mL
ChEBI CHEBI:26708
Grade ICP/ICP-MS
Chemical Name or Material Sodium Standard
Show More Show Less
compliance-icons
Product Identifier
  • 1000 µg/mL Sodium
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: nitric acid, sodiumfrom sodium carbonate

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.