missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiocyanate solution 0.05mol/L, Fisher Chemical™
BIOPHARMA

Product Code. 12973624 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12973624 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12973624 Supplier Fisher Chemical Supplier No. VL 310 1000
  • ONLINE EXCLUSIVE PRICE
    63.25 €
    51.35 € / 1000mL
    Save 11.90 €
    18% Off
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Bottle
CAS 57-55-6
Molecular Formula C3H8O2
Concentration 0.05 M
InChI Key DNIAPMSPPWPWGF-UHFFFAOYSA-N
SMILES CC(CO)O
IUPAC Name propane-1,2-diol
Molecular Weight (g/mol) 76.095
PubChem CID 1030
Quantity 1000 mL
ChEBI CHEBI:16997
Chemical Name or Material Sodium Thiosulphate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.