missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate, Acculute Standard Volumetric Solution, Final Concentration 0.1N

Product Code. 11379329
Change view
Click to view available options
Quantity:
1 U
6 U
Unit Size:
1 unit
6 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11379329 6 U 6 units
11369329 1 U 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11379329 Supplier Thermo Scientific Alfa Aesar Supplier No. 088628.LS
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7772-98-7 | Na2O3S2 | 158.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7772-98-7
Molecular Formula Na2O3S2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00003499
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
Synonym sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
PubChem CID 24477
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Linear Formula Na2S2O3
IUPAC Name disodium sulfanidesulfonate
Formula Weight 158.1
Quantity 6 U
Physical Form Liquid
Chemical Name or Material Sodium thiosulfate Acculute Standard Volumetric Solution, Final Concentration 0.1N
What are the instructions for dilution of 1 unit of Sodium thiosulfate, Acculute Standard Volumetric Solution, Final Concentration 0.1N (Cat. No. 088628.LS, 088628.LQ)?

The following is the dilution protocol:
- Transfer entire contents of 1 unit bottle into a 1L volumetric flask.
- Rinse bottle residue, the threads of the bottle, and inner bottle cap each with DI water, and transfer to the 1L flask.
- Repeat rinse three times. Bring the 1L flask to volume with DI water and mix well.
The final solution in the 1L flask has a concentration of 0.1N Sodium thiosulfate.

What is 1 unit of Sodium thiosulfate, Acculute Standard Volumetric Solution, Final Concentration 0.1N (Cat. No. 088628.LS, 088628.LQ)?

1 unit of Sodium thiosulfate, Acculute Standard Volumetric Solution, Final Concentration 0.1N, when diluted to 1 L, will yield a final concentration of 0.1N Sodium thiosulfate solution.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.