missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulphate 1.0 M (1.0 N) Analytical Volumetric Solution (AVL), Reagecon
Shop All Reagecon Products

Description
- Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
- Accurate, stable, reproducible and fully factorized.
- Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
- Stability is proven (either opened or unopened) for the shelf life of this product.
- Certified by Reagecon’s ISO 17025 Accredited Test Method (INAB Ref: 264T).
- Certified to a specification of ± 0.1%.
- Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
- May be used for manual titrations, or automatic titrations, to determine the end points by detection of any of several properties.
- Oxidation/reduction titrations depend on changes in the electropotential of the oxidation reduction system and always involves a reaction between an oxidizing and reducing pair.
Specifications
Specifications
| Description | Accuracy ± 0.01% |
| Packaging | HDPE Bottle GL45 |
| Physical Form | Liquid |
| Concentration | 1.0 M |
| Technique | Titration |
| Quantity | 1 L |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?