Learn More
Memmert™ Software AtmoCONTROL for SF160plus
Product Code. 17528165 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
SF160plus Sterilizer
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
17528165 SF160plus Sterilizer Each
1 options
Product Code. 17528165 Supplier Memmert™ Supplier No. SF160plusFDAQ1
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
AtmoCONTROL Software meets the requirements for the use of electronically stored data sets and electronic signatures.
  • Software conforming to FDA AtmoCONTROL
  • Meets the requirements for the use of electronically stored data sets and electronic signatures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
  • Base licence for the control of one unit
  • Respective IQ/OQ documents available in German and English language (without surcharge)
Specifications
Certifications/Compliance Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
For Use With (Equipment) SF160plus Sterilizer
Product Type Software