Learn More
Solution Kit of References B, 9x100mL solutions EU Pharmacopoeia, Fisher Chemical™
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Brand: Fisher Chemical OP 010 100
Code : 04
Additional Details : Weight : 0.90000kg
Specifications
Solution Kit of References B | |
9 x 100 mL | |
Liquid |
Bottle | |
Eu. Ph. |
Safety and Handling
- Danger
Hazard Category
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 2
- Reproductive toxicity Category 1B
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Carcinogenicity Category 1B
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H360-May damage fertility or the unborn child.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands, forearms andface thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : hydrochloric acid, cobalt(II) chloride,hexahydrate, copper(II) sulphate, pentahydrate