missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Somatostatin Antibody (7G5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
196.00 € - 600.00 €
Specifications
| Antigen | Somatostatin |
|---|---|
| Clone | 7G5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18725003
|
Novus Biologicals
NBP2-37447 |
0.1 mL |
600.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653936
|
Novus Biologicals
NBP2-37447-0.025ml |
0.025 mL |
196.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Somatostatin Monoclonal specifically detects Somatostatin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence, CyTOF-ready.Specifications
| Somatostatin | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| RUO | |
| Human | |
| Growth hormone release-inhibiting factor, SMST, somatostatin, somatostatin-14, somatostatin-28 | |
| SST | |
| IgG1 | |
| Affinity Purified |
| 7G5 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Breast Cancer, Cancer, Stem Cell Markers | |
| PBS with 0.05% Sodium Azide | |
| 6750 | |
| Purified recombinant fragment of human SST (AA: 1-116) expressed in E. Coli. | |
| Primary | |
| 12.7 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title