missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Somatostatin R2/SSTR2 Antibody (SR1695), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21663-100ul
This item is not returnable.
View return policy
Description
Somatostatin R2/SSTR2 Monoclonal antibody specifically detects Somatostatin R2/SSTR2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
Specifications
| Somatostatin R2/SSTR2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| SR1695 | |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
| somatostatin receptor 2, somatostatin receptor type 2, SRIF-1, SS2R, SS-2-R, SS2-R | |
| A synthesized peptide derived from human Somatostatin R2/SSTR2 (Uniprot #: P30874) | |
| 100 μg | |
| Alzheimers Research, Cancer, Cell Cycle and Replication, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 6752 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction