Learn More
Sortilin Antibody (CL6526) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76498B
Product Code. 30297910
  • 737.00 € / 0.10mL
Description
Sortilin Monoclonal antibody specifically detects Sortilin in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sortilin
Monoclonal
Biotin
100 kDa NT receptor, Glycoprotein 95, Gp95LDLCQ6, Neurotensin receptor 3, NT3NTR3, sortilin, sortilin 1
This antibody was developed using a recombinant protein derived from Q99523, with the exact immunogen sequence remaining proprietary.
0.1 mL
Golgi Apparatus Markers
6272
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence
CL6526
PBS
Mouse
Protein A purified
RUO
Primary
Human, Mouse
Purified