Learn More
Sortilin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NBP1-89745
 View more versions of this product
Product Code. 18745093
  • 529.00 € / 0.10mL
Description
Sortilin Polyclonal specifically detects Sortilin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
0.1 mL
IgG
This antibody was developed against Recombinant Protein corresponding to amino acids:LERNCEEKDYTIWLAHSTDPEDYEDGCILGYKEQFLRLRKSSVCQNGRDYVVTKQPSICLCSLEDFLCDFGYYRPENDSKCVEQPELKGHDLEFCLYGREEHLTTNGYRKIPGDKCQGGVNPVREVKD
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.