missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sorting Nexin 31 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06489-50ul
This item is not returnable.
View return policy
Description
Sorting Nexin 31 Polyclonal antibody specifically detects Sorting Nexin 31 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Sorting Nexin 31 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 | |
| MGC39715, sorting nexin 31, sorting nexin-31 | |
| Produced in rabbits immunized with E. coli-derived Human Sorting Nexin 31 fragment. | |
| 50 μg | |
| Signal Transduction | |
| 169166 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction