missing translation for 'onlineSavingsMsg'
Learn More

Sotalol hydrochloride, 98%

Product Code. 15495939
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495939 5 g 5g
15405949 100 mg 100mg
15485939 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15495939 Supplier Thermo Scientific Alfa Aesar Supplier No. J63772.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent beta adrenergic antagonist | CAS: 959-24-0 | C12H21ClN2O3S | 308.82 g/mol

Sotalol hydrochloride is used as a potent beta adrenergic antagonist, prolongs the action potential and increases the refractory period. Sotalol hydrochloride is also considered a non-selective β blocker and a potassium channel blocker with an IC50 of 43 μM.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sotalol hydrochloride is used as a potent beta adrenergic antagonist, prolongs the action potential and increases the refractory period. Sotalol hydrochloride is also considered a non-selective β blocker and a potassium channel blocker with an IC50 of 43 µM.

Solubility
Soluble in phosphate buffered saline, DMSO, ethanol, water, and methanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 959-24-0
Molecular Formula C12H21ClN2O3S
Molecular Weight (g/mol) 308.82
MDL Number MFCD00242937
InChI Key VIDRYROWYFWGSY-UHFFFAOYNA-N
Synonym sotalol hydrochloride, sotalol hcl, betapace, betapace af, sotacor, sotalex, berlex, mead johnson 1999, sorine
PubChem CID 66245
ChEBI CHEBI:9207
IUPAC Name hydrogen N-(4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}phenyl)methanesulfonamide chloride
SMILES [H+].[Cl-].CC(C)NCC(O)C1=CC=C(NS(C)(=O)=O)C=C1

Specifications

Color White
Quantity 5 g
Merck Index 14,8728
Solubility Information Soluble in phosphate buffered saline,DMSO,ethanol,water,and methanol.
Formula Weight 308.82
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Sotalol hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.