missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX14 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93022-0.02ml
This item is not returnable.
View return policy
Description
SOX14 Polyclonal antibody specifically detects SOX14 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| SOX14 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| HMG box transcription factor SOX-14, MGC119898, MGC119899, SRY (sex determining region Y)-box 14, SRY-box 14, transcription factor SOX-14 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 91-240 of human SOX14 (NP_004180.1). RYVFPLPYLGDTDPLKAAGLPVGASDGLLSAPEKARAFLPPASAPYSLLDPAQFSSSAIQKMGEVPHTLATGALPYASTLGYQNGAFGSLSCPSQHTHTHPSPTNPGYVVPCNCTAWSASTLQPPVAYILFPGMTKTGIDPYSSAHATAM | |
| 0.02 mL | |
| Neuroscience | |
| 8403 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction