Learn More
SOX21 Antibody (CL4688) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-59059PE
Product Code. 30298121
  • 737.00 € / 0.10mL
Description
SOX21 Monoclonal antibody specifically detects SOX21 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SOX21
Monoclonal
PE
SOX25SOX-A, SOXA, SRY (sex determining region Y)-box 21, SRY-box 21, transcription factor SOX-21
This antibody was developed using a synthetic peptide derived from Q9Y651, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human, Mouse
Purified
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL4688
PBS
Mouse
Protein A purified
RUO
11166
Store at 4⪚C in the dark.
IgG2a